accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClNOO

CAS RN 610-40-2

IUPAC Name: 4-chloro-1,2-dinitrobenzene
Synonyms: 1,2-DINO2 4-CL benzene
Molecular Weight (g/mol): 202.55
Molecular Formula: C6H3ClN2O4
InChi Key: QVQSOXMXXFZAKU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)