accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610-97-9

IUPAC Name: methyl 2-iodobenzoate
Synonyms: methyl 2-iodobenzoate
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: BXXLTVBTDZXPTN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1I
Boiling Point 
  • (1)
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)