accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO

CAS RN 610-99-1

IUPAC Name: 1-(2-hydroxyphenyl)propan-1-one
Synonyms: O-hydroxypropiophenone
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: KDUWXMIHHIVXER-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=C1O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)