accessibility menu, dialog, popup

UserName

Viewing profile as user:
KHOOOHOHOOOO

CAS RN 6100-20-5

IUPAC Name: potassium oxalic acid hydrogen oxalate
Synonyms: potassium oxalic acid hydrogen oxalate
Molecular Weight (g/mol): 218.16
Molecular Formula: C4H3KO8
InChi Key: GANDVAJEIJXBQJ-UHFFFAOYSA-M
SMILES: [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (2)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)