accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OClClH3CNCH3H3COOOONCH3CH3H3C

CAS RN 6101-15-1

IUPAC Name: trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dihydrate dichloride
Synonyms: suxamethonium chloride 2H2O succinyldicholine dichloride dihydrate succinylcholine dichloride dihydrate suxamethonium chloride dihydrate succinylcholine chloride dihydrate succinylcholine chloride 2H2O
Molecular Weight (g/mol): 397.33
Molecular Formula: C14H34Cl2N2O6
InChi Key: FFSBEIRFVXGRPR-UHFFFAOYSA-L
SMILES: O.O.[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)