accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3(S)(S)(S)O(R)OOOHONOO

CAS RN 6106-46-3

IUPAC Name: (1S,2R,4S,5S)-7-[(3-hydroxy-2-phenylpropanoyl)oxy]-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium nitrate
Synonyms: (1S,2R,4S,5S)-7-[(3-hydroxy-2-phenylpropanoyl)oxy]-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium nitrate
Molecular Weight (g/mol): 380.4
Molecular Formula: C18H24N2O7
InChi Key: BSQIVYOSLFLSGE-ODDNLWGVSA-N
SMILES: [O-][N+]([O-])=O.C[N+]1(C)[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)C(CO)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results