accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610763-72-9

IUPAC Name: methyl 4-(acetyloxy)-2-{[3-(benzyloxy)-6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Synonyms: methyl 4-(acetyloxy)-2-{[3-(benzyloxy)-6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Molecular Weight (g/mol): 1073.31
Molecular Formula: C48H56Cl3NO20
InChi Key: HNINFVMNWIPTPG-UHFFFAOYNA-N
SMILES: COC(=O)C1(CC(OC(C)=O)C(NC(=O)OCC(Cl)(Cl)Cl)C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O)OC1C(O)C(COCC2=CC=CC=C2)OC(OC2=CC=C(OC)C=C2)C1OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results