accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiOO

CAS RN 6108-23-2

IUPAC Name: lithium(1+) formate
Synonyms: lithium formate
Molecular Weight (g/mol): 51.96
Molecular Formula: CHLiO2
InChi Key: XKPJKVVZOOEMPK-UHFFFAOYSA-M
SMILES: [Li+].[O-]C=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results