accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6109-35-9

IUPAC Name: 4-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Synonyms: 4-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Molecular Weight (g/mol): 245.75
Molecular Formula: C15H16ClN
InChi Key: YYCOJKPDBCMVPP-UHFFFAOYNA-N
SMILES: Cl.C1NCC2=CC=CC=C2C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results