accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-06-3

IUPAC Name: 2,4-dichloro-1-nitrobenzene
Synonyms: 2,4-dichloronitrobenzene
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: QUIMTLZDMCNYGY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results