accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C

CAS RN 611-14-3

IUPAC Name: 1-ethyl-2-methylbenzene
Synonyms: o-ethyltoluene 1,2-methylethylbenzene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: HYFLWBNQFMXCPA-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)