accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHHO

CAS RN 611-24-5

IUPAC Name: 2-(methylamino)phenol
Synonyms: 2-(methylamino)phenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: JHKKTXXMAQLGJB-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results