accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-69-8

IUPAC Name: 2-methyl-1-phenylpropan-1-ol
Synonyms: isopropyl benzyl alcohol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: GMDYDZMQHRTHJA-UHFFFAOYNA-N
SMILES: CC(C)C(O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results