accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-70-1

IUPAC Name: 2-methyl-1-phenylpropan-1-one
Synonyms: 2-methyl-1-phenylpropan-1-one
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: BSMGLVDZZMBWQB-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)