CAS RN 611-71-2
(R)-(-)-Mandelic acid, 99%, Thermo Scientific Chemicals
CAS: 611-71-2 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00064251 InChI Key: IWYDHOAUDWTVEP-SSDOTTSWSA-N Synonym: r-mandelic acid,r---mandelic acid,d-mandelic acid,r-2-hydroxy-2-phenylacetic acid,--mandelic acid,d--mandelic acid,d---mandelic acid,2r-2-hydroxy-2-phenylacetic acid,r-alpha-hydroxyphenylacetic acid,d-2-phenylglycolic acid PubChem CID: 11914 ChEBI: CHEBI:17656 IUPAC Name: (2R)-2-hydroxy-2-phenylacetic acid SMILES: O[C@@H](C(O)=O)C1=CC=CC=C1
(R)-(-)-Mandelic acid, 98%
CAS: 611-71-2 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00064251 InChI Key: IWYDHOAUDWTVEP-SSDOTTSWSA-N Synonym: r-mandelic acid,r---mandelic acid,d-mandelic acid,r-2-hydroxy-2-phenylacetic acid,#NAME?,d--mandelic acid,d---mandelic acid,2r-2-hydroxy-2-phenylacetic acid,r-alpha-hydroxyphenylacetic acid,d-2-phenylglycolic acid PubChem CID: 11914 ChEBI: CHEBI:17656 IUPAC Name: (2R)-2-hydroxy-2-phenylacetic acid SMILES: O[C@@H](C(O)=O)C1=CC=CC=C1
D-(-)-Mandelic Acid 99.0+%, TCI America™
CAS: 611-71-2 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00064251 InChI Key: IWYDHOAUDWTVEP-SSDOTTSWSA-N Synonym: r-mandelic acid,r---mandelic acid,d-mandelic acid,r-2-hydroxy-2-phenylacetic acid,#NAME?,d--mandelic acid,d---mandelic acid,2r-2-hydroxy-2-phenylacetic acid,r-alpha-hydroxyphenylacetic acid,d-2-phenylglycolic acid PubChem CID: 11914 ChEBI: CHEBI:17656 IUPAC Name: (2R)-2-hydroxy-2-phenylacetic acid SMILES: O[C@@H](C(O)=O)C1=CC=CC=C1
Sigma Aldrich Thiol-Dpeg™4-Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich ™-(-)-Mandelic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Gly-Ala
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%
CAS: 611-71-2 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00064251 InChI Key: IWYDHOAUDWTVEP-SSDOTTSWSA-N Synonym: r-mandelic acid,r---mandelic acid,d-mandelic acid,r-2-hydroxy-2-phenylacetic acid,#NAME?,d--mandelic acid,d---mandelic acid,2r-2-hydroxy-2-phenylacetic acid,r-alpha-hydroxyphenylacetic acid,d-2-phenylglycolic acid PubChem CID: 11914 ChEBI: CHEBI:17656 IUPAC Name: (2R)-2-hydroxy-2-phenylacetic acid SMILES: O[C@@H](C(O)=O)C1=CC=CC=C1