accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 611-73-4

IUPAC Name: 2-oxo-2-phenylacetic acid
Synonyms: benzoylformic acid phenylgloxylic acid phenylglyoxalate calcium salt phenylglyoxylic acid phenylglyoxylate (substance) potassium salt phenylglyoxylic acid sodium salt phenylglyoxylic acid phenylglyoxilic acid α-oxobenzeneacetic acid
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: FAQJJMHZNSSFSM-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (7)
  • (2)
Quantity 
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
Physical Form 
  • (4)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (6)
  • (2)
  • (1)
  • (7)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)