accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-17-9

IUPAC Name: 1,4-dihydronaphthalene
Synonyms: 1,4-dihydronaphthalene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: FUPIVZHYVSCYLX-UHFFFAOYSA-N
SMILES: C1C=CCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results