accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-25-9

IUPAC Name: (2-nitrophenyl)methanol
Synonyms: 2-nitrobenzyl alcohol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: BWRBVBFLFQKBPT-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)