accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO(E)

CAS RN 612-40-8

IUPAC Name: 2-[(1E)-2-carboxyeth-1-en-1-yl]benzoic acid
Synonyms: 2-[(1E)-2-carboxyeth-1-en-1-yl]benzoic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: SCWPNMHQRGNQHH-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results