accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-75-9

IUPAC Name: 3,3'-dimethyl-1,1'-biphenyl
Synonyms: 3,3'-dimethylbiphenyl
Molecular Weight (g/mol): 182.27
Molecular Formula: C14H14
InChi Key: GVEDOIATHPCYGS-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)C1=CC(C)=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)