CAS RN 612-82-8
o-Tolidine dihydrochloride, 99%
CAS: 612-82-8 Molecular Formula: C14H16N2·2HCl Molecular Weight (g/mol): 285.2 InChI Key: LUKPNZHXJRJBAN-UHFFFAOYSA-N Synonym: 3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl PubChem CID: 108938 IUPAC Name: 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride SMILES: CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
o-Tolidine dihydrochloride, 97%
CAS: 612-82-8 Molecular Formula: C14H18Cl2N2 Molecular Weight (g/mol): 285.212 MDL Number: MFCD00012960 InChI Key: LUKPNZHXJRJBAN-UHFFFAOYSA-N Synonym: 3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl PubChem CID: 108938 IUPAC Name: 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride SMILES: CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
o-Tolidine dihydrochloride, ACS
CAS: 612-82-8 Molecular Formula: C14H18Cl2N2 Molecular Weight (g/mol): 285.212 MDL Number: MFCD00012960 InChI Key: LUKPNZHXJRJBAN-UHFFFAOYSA-N Synonym: 3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl PubChem CID: 108938 IUPAC Name: 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride SMILES: CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
o-Tolidine Dihydrochloride 98.0+%, TCI America™
CAS: 612-82-8 Molecular Formula: C14H18Cl2N2 Molecular Weight (g/mol): 285.212 MDL Number: MFCD00012960 InChI Key: LUKPNZHXJRJBAN-UHFFFAOYSA-N Synonym: 3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl PubChem CID: 108938 IUPAC Name: 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride SMILES: CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl