accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 6120-95-2

IUPAC Name: 1-phenylcyclopropane-1-carboxylic acid
Synonyms: 1-phenylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: IWWCCNVRNHTGLV-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)