accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CH3CCH3PHCH3CH3H3C

CAS RN 612088-55-8

IUPAC Name: di-tert-butyl(phenyl)phosphanium; tetrafluoroboranuide
Synonyms: di-tert-butyl(phenyl)phosphanium tetrafluoroborate
Molecular Weight (g/mol): 310.12
Molecular Formula: C14H24BF4P
InChi Key: HRDPEVWZXUWEFR-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.CC(C)(C)[PH+](C1=CC=CC=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results