accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6125-57-1

IUPAC Name: methyl 4-hydroxycyclohexane-1-carboxylate
Synonyms: methyl 4-hydroxycyclohexane-1-carboxylate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: HYDYVXROZHFTGB-UHFFFAOYSA-N
SMILES: COC(=O)C1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results