accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61266-70-4

IUPAC Name: (oxetan-2-yl)methanol
Synonyms: oxetan-2-ylmethanol
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: PQZJTHGEFIQMCO-UHFFFAOYNA-N
SMILES: OCC1CCO1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)