CAS RN 613-06-9
IUPAC Name:
2,3-dimethylanthracene
Synonyms:
2,3-dimethylanthracene
Molecular Weight (g/mol):
206.29
Molecular Formula:
C16H14
InChi Key:
OGVRJXPGSVLDRD-UHFFFAOYSA-N
SMILES:
CC1=C(C)C=C2C=C3C=CC=CC3=CC2=C1
1
–
1
of
1
results
1
2,3-Dimethylanthracene 98.0+%, TCI America™
CAS: 613-06-9 Molecular Formula: C16H14 Molecular Weight (g/mol): 206.288 InChI Key: OGVRJXPGSVLDRD-UHFFFAOYSA-N PubChem CID: 69170 IUPAC Name: 2,3-dimethylanthracene SMILES: CC1=CC2=CC3=CC=CC=C3C=C2C=C1C