accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 613-12-7

IUPAC Name: 2-methylanthracene
Synonyms: 2-methylanthracene
Molecular Weight (g/mol): 192.26
Molecular Formula: C15H12
InChi Key: GYMFBYTZOGMSQJ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C3C=CC=CC3=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results