accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3

CAS RN 613-33-2

IUPAC Name: 4,4'-dimethyl-1,1'-biphenyl
Synonyms: 4,4'-dimethyl-1,1'-biphenyl
Molecular Weight (g/mol): 182.27
Molecular Formula: C14H14
InChi Key: RZTDESRVPFKCBH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)