accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 613-37-6

IUPAC Name: 4-methoxy-1,1'-biphenyl
Synonyms: 4-methoxybiphenyl
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: RHDYQUZYHZWTCI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)