accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 613-69-4

IUPAC Name: 2-ethoxybenzaldehyde
Synonyms: benzaldehyde, 2-ethoxy-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: DUVJMSPTZMCSTQ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C=O
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)