accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613-80-9

IUPAC Name: 2-(naphthalen-2-yloxy)naphthalene
Synonyms: bis(2-naphthyl)ether
Molecular Weight (g/mol): 270.33
Molecular Formula: C20H14O
InChi Key: DZRLNYVDCIYXPG-UHFFFAOYSA-N
SMILES: O(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)