accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 613-94-5

IUPAC Name: benzohydrazide
Synonyms: benzohydrazide hydrazide benzoic acid C6H5-CO-NH-NH2 hydrazid kyseliny benzoove
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: WARCRYXKINZHGQ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (4)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (7)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)