accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6130-75-2

IUPAC Name: 1,2,4-trichloro-5-methoxybenzene
Synonyms: 2,4,5-trichloroanisole
Molecular Weight (g/mol): 211.47
Molecular Formula: C7H5Cl3O
InChi Key: SXKBHOQOOGRFJF-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results