accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OH2OH2OH2OH2ONaNaOOO

CAS RN 6132-02-1

IUPAC Name: disodium decahydrate carbonate
Synonyms: disodium decahydrate carbonate
Molecular Weight (g/mol): 286.14
Molecular Formula: CH20Na2O13
InChi Key: XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
Grade 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)