accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNFFF

CAS RN 613239-75-1

IUPAC Name: {4-[5-(trifluoromethyl)pyridin-2-yl]phenyl}methanol
Synonyms: {4-[5-(trifluoromethyl)pyridin-2-yl]phenyl}methanol
Molecular Weight (g/mol): 253.22
Molecular Formula: C13H10F3NO
InChi Key: VRVOASJNEFKGGI-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results