accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61324-93-4

IUPAC Name: 1-fluoro-4-methoxy-2-nitrobenzene
Synonyms: 1-fluoro-4-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 171.13
Molecular Formula: C7H6FNO3
InChi Key: ZRIKJXDEJYMBEJ-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results