accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOFFFF

CAS RN 61324-97-8

IUPAC Name: 2-fluoro-1-nitro-3-(trifluoromethyl)benzene
Synonyms: 2-fluoro-1-nitro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: PGKWPKDZNHZBQK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results