accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrBr

CAS RN 61326-44-1

IUPAC Name: 1-bromo-4-[1,2,2-tris(4-bromophenyl)ethenyl]benzene
Synonyms: 1-bromo-4-[1,2,2-tris(4-bromophenyl)ethenyl]benzene
Molecular Weight (g/mol): 648.03
Molecular Formula: C26H16Br4
InChi Key: BIRLDGKMJJEZRI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(=C(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results