accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61336-70-7

IUPAC Name: (2S,5R,6R)-6-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Synonyms: trimox zimox larotid moxaline polymox amoxipen robamox
Molecular Weight (g/mol): 419.45
Molecular Formula: C16H25N3O8S
InChi Key: MQXQVCLAUDMCEF-CWLIKTDRSA-N
SMILES: O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=C(O)C=C3)C(=O)N2[C@H]1C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Grade 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Certifications/Compliance 
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)