CAS RN 6136-37-4
CAS RN 6136-37-4
IUPAC Name:
1-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms:
1-methylxanthine
Molecular Weight (g/mol):
166.14
Molecular Formula:
C6H6N4O2
InChi Key:
MVOYJPOZRLFTCP-UHFFFAOYSA-N
SMILES:
CN1C(=O)NC2=C(NC=N2)C1=O
1
–
1
of
1
results
1
1-Methylxanthine 97.0+%, TCI America™

CAS: 6136-37-4 Molecular Formula: C6H6N4O2 Molecular Weight (g/mol): 166.14 MDL Number: MFCD00005561 InChI Key: MVOYJPOZRLFTCP-UHFFFAOYSA-N PubChem CID: 80220 ChEBI: CHEBI:68444 IUPAC Name: 1-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione SMILES: CN1C(=O)NC2=C(NC=N2)C1=O