accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61366-43-6

IUPAC Name: 2-amino-3-(benzyloxy)propan-1-ol hydrochloride
Synonyms: 2-amino-3-(benzyloxy)propan-1-ol hydrochloride
Molecular Weight (g/mol): 217.69
Molecular Formula: C10H16ClNO2
InChi Key: BJWHQBTUHVIRJJ-UHFFFAOYNA-N
SMILES: Cl.NC(CO)COCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results