accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NN

CAS RN 61379-64-4

IUPAC Name: 4-cyclopentylpiperazin-1-amine
Synonyms: 4-cyclopentylpiperazin-1-amine
Molecular Weight (g/mol): 169.27
Molecular Formula: C9H19N3
InChi Key: QYHRIASMJNLWHJ-UHFFFAOYSA-N
SMILES: NN1CCN(CC1)C1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results