accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3OOCH3

CAS RN 6138-26-7

IUPAC Name: 1,2,3-trimethyl propane-1,2,3-tricarboxylate
Synonyms: 1,2,3-trimethyl propane-1,2,3-tricarboxylate
Molecular Weight (g/mol): 218.21
Molecular Formula: C9H14O6
InChi Key: IXINWTSBOAMACD-UHFFFAOYSA-N
SMILES: COC(=O)CC(CC(=O)OC)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results