accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 614-14-2

IUPAC Name: 1-phenylbutan-1-ol
Synonyms: 1-phenylbutan-1-ol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: HQRWWHIETAKIMO-UHFFFAOYNA-N
SMILES: CCCC(O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results