accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOO

CAS RN 614-19-7

IUPAC Name: 3-amino-3-phenylpropanoic acid
Synonyms: 3-amino-3-phenylpropanoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: UJOYFRCOTPUKAK-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)