accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-45-9

IUPAC Name: tert-butyl benzenecarboperoxoate
Synonyms: T-butyl benzoyl peroxide benzoyl T-butyl peroxide
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: GJBRNHKUVLOCEB-UHFFFAOYSA-N
SMILES: CC(C)(C)OOC(=O)C1=CC=CC=C1
Boiling Point 
  • (7)
  • (2)
Percent Purity 
  • (1)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (5)
  • (7)
  • (2)
  • (1)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)