accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-77-7

IUPAC Name: (2-methylphenyl)urea
Synonyms: urea, (2-methylphenyl)-
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: BLSVCHHBHKGCSQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results