accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-83-5

CH3ONHCH3Br

CAS RN 614-83-5

IUPAC Name: N-(2-bromo-4-methylphenyl)acetamide
Synonyms: N-(2-bromo-4-methylphenyl)acetamide
Molecular Weight (g/mol): 228.09
Molecular Formula: C9H10BrNO
InChi Key: UUDGTWKIIUEVJD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results