accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HN

CAS RN 614-96-0

IUPAC Name: 5-methyl-1H-indole
Synonyms: 5-methylindole
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: YPKBCLZFIYBSHK-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)