accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 614-98-2

IUPAC Name: ethyl furan-3-carboxylate
Synonyms: ethyl furan-3-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: LOFDXZJSDVCYAS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=COC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)